Novus Biologicals
Manufacturer Code:NBP288830
Catalog # NB036450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-EARQSPPLNRFD from the internal region (near C Terminus) of the protein sequence according to NP_038490.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRB3R adrenergic beta-3- receptor B3AR beta-3 adrenergic receptor Beta-3 adrenoceptor Beta-3 adrenoreceptor BETA3AR FLJ99960; adrenergic, beta-3-, receptor; Beta-3 adrenergic receptor; Beta-3 adrenoceptor; Beta-3 adrenoreceptor
Gene Aliases: ADRB3; ADRB3R; B3AR; BETA3AR
UniProt ID: (Human) P13945
Entrez Gene ID: (Human) 155
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.