Novus Biologicals
Manufacturer Code:NBP290959UV
Catalog # NB041444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 110 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human beta 2-Microglobulin/FCGRT (P55899 & P61769 Met1-Ser297 & Met1-Met119). |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-chain beta chain of MHC class I molecules beta-2-microglobin beta-2-microglobulin Fc fragment of IgG receptor transporter alpha FcRn FcRn alpha chain heavy chain of the major histocompatibility complex class I-like Fc receptor IgG Fc fragment receptor transporter alpha chain IgG receptor FcRn large subunit p51 IMD43 immunoglobulin receptor intestinal heavy chain major histocompatibility complex class I-like Fc receptor Neonatal Fc receptor neonatal Fc-receptor for Ig transmembrane alpha chain of the neonatal receptor; beta chain of MHC class I molecules; beta-2-microglobin; Beta-2-microglobulin; Beta-2-microglobulin form pI 5.3
Gene Aliases: B2M; CDABP0092; HDCMA22P; IMD43
UniProt ID: (Human) P61769
Entrez Gene ID: (Human) 567
Molecular Function:
defense/immunity protein
major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.