Invitrogen
Manufacturer Code:PA514118
Catalog # PIPA514118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 345-373 amino acids from human ADRB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; beta 2-AR; Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; Beta-2 adrenoreceptor Beta-2 adrenoceptor Beta-2 adrenergic receptor ADRB2 ADRB2R B2AR; catecholamine receptor
Gene Aliases: Adrb-2; ADRB2; ADRB2R; ADRBR; B2AR; Badm; BAR; BETA2AR; Gpcr7
UniProt ID: (Human) P07550, (Mouse) Q8BH38
Entrez Gene ID: (Human) 154, (Mouse) 11555
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.