Invitrogen
Manufacturer Code:PA125659
Catalog # PIPA125659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 335-408 of human beta 2 Adrenergic Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRB2R ADRBR B2AR BAR BETA2AR; adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; catecholamine receptor
Gene Aliases: ADRB2; ADRB2R; ADRBR; B2AR; BAR; BETA2AR
UniProt ID: (Human) P07550
Entrez Gene ID: (Human) 154
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.