R&D Systems
Manufacturer Code:MAB100401SP
Catalog # MAB100401SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 586107 |
Immunogen | NS0 mouse myeloma cell line transfected with human beta-2 Adrenergic R/ADRB2 Accession No. P07550 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Mouse Monoclonal Antibody
Protein Aliases: ADRB2 ADRB2R ADRBR adrenergic beta-2- receptor surface B2AR B2ARADRB2R BAR beta-2 adrenergic receptor beta2 AdrenergicR beta-2 AdrenergicR Beta-2 adrenoceptor Beta-2 adrenoreceptor BETA2AR catecholamine receptor; adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; catecholamine receptor
Gene Aliases: ADRB2; ADRB2R; ADRBR; B2AR; BAR; BETA2AR
UniProt ID: (Human) P07550
Entrez Gene ID: (Human) 154
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.