Novus Biologicals
Manufacturer Code:NB12013989
Catalog # NB12013989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide: DFRIAFQELLC LRRSSLKAYGN GYSSNGNTGE QSGYHVEQEKE NKLLCEDLPGT EDFVGHQGTVP SDNIDSQGRNCS T corresponding to amino acids 335-408 of Human Beta 2 Adrenergic Receptor (NP_000015.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRBR adrenergic beta-2- receptor surface B2ARADRB2R BAR beta-2 adrenergic receptor Beta-2 adrenoceptor Beta-2 adrenoreceptor BETA2AR catecholamine receptor; adrenergic, beta-2-, receptor, surface; adrenoceptor beta 2 surface; adrenoceptor beta 2, surface; Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; catecholamine receptor
Gene Aliases: ADRB2; ADRB2R; ADRBR; B2AR; BAR; BETA2AR
UniProt ID: (Human) P07550
Entrez Gene ID: (Human) 154
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.