Novus Biologicals
Manufacturer Code:NBP299973AF700
Catalog # NB063802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant beta-13-Glucuronyltransferase 3/B3GAT3 (NP_036332.2 Glu72-Val335). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); Beta-13-glucuronyltransferase 3 beta-13-glucuronyltransferase 3 (glucuronosyltransferase I) EC 2.4.1.135 galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3 GlcAT-I GLCATI GlcUAT-I Glucuronosyltransferase I Sqv-8-like protein UDP-GlcUA:Gal beta-13-Gal-R glucuronyltransferase; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GlcAT-I; GlcUAT-I; Glucuronosyltransferase I; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase
Gene Aliases: B3GAT3; GLCATI; glcUAT-I; JDSCD
UniProt ID: (Human) O94766
Entrez Gene ID: (Human) 26229
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.