Novus Biologicals
Manufacturer Code:NBP2443520.2MG
Catalog # NBP24435202
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NK-1 |
Immunogen | Human peripheral blood mononuclear cells |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-glucuronyltransferase 1; Beta-13-glucuronyltransferase 1 beta-13-glucuronyltransferase 1 (glucuronosyltransferase P) EC 2.4.1.135 galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1 GlcAT-P GLCATPGLCUATP glcUAT-P Glucuronosyltransferase P NK1 NK-1 UDP-GlcUA:glycoprotein beta-13-glucuronyltransferase; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GlcAT-P; GlcUAT-P; Glucuronosyltransferase P; LEU7 antigen; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Aliases: B3GAT1; CD57; GLCATP; GLCUATP; HNK1; LEU7; NK-1; NK1
UniProt ID: (Human) Q9P2W7
Entrez Gene ID: (Human) 27087
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.