Novus Biologicals
Manufacturer Code:NBP119788V
Catalog # NBP119788V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an N-terminal portion of the human B3TGAT1 protein (between residues 1-50) [UniProt Q9P2W7] |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1,3-glucuronyltransferase 1; Beta-13-glucuronyltransferase 1 beta-13-glucuronyltransferase 1 (glucuronosyltransferase P) EC 2.4.1.135 galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1 GlcAT-P GLCATPGLCUATP glcUAT-P Glucuronosyltransferase P NK1 NK-1 UDP-GlcUA:glycoprotein beta-13-glucuronyltransferase; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GlcAT-P; GlcUAT-P; Glucuronosyltransferase P; LEU7 antigen; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Aliases: B3GAT1; CD57; GLCATP; GLCUATP; HNK1; LEU7; NK-1; NK1
UniProt ID: (Human) Q9P2W7
Entrez Gene ID: (Human) 27087
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.