Invitrogen
Manufacturer Code:PA5113632
Catalog # PIPA5113632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ADRB1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adrb1 Adrb-1 Adrb1r adrenergic receptor beta 1 adrenergic beta-1- receptor adrenoceptor beta 1 B1AR beta 1-adrenergic receptor beta 1-AR beta 1-AR Beta-1 adrenergic receptor Beta-1 adrenoceptor Beta-1 adrenoreceptor BETA1AR beta-AR cardiac beta adrenergic receptor RATB1AR RHR; adrenergic, beta-1-, receptor; Beta-1 adrenergic receptor; Beta-1 adrenoceptor; Beta-1 adrenoreceptor
Gene Aliases: ADRB1; ADRB1R; B1AR; BETA1AR; RHR
UniProt ID: (Human) P08588
Entrez Gene ID: (Human) 153
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.