Novus Biologicals
Manufacturer Code:NB600978
Catalog # NB600978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
In vitro Assay (IV) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence ESDEARRCYNDPK corresponding to internal region according to NP_000675.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRB1RRHR adrenergic beta-1- receptor B1AR beta-1 adrenergic receptor Beta-1 adrenoceptor Beta-1 adrenoreceptor BETA1AR; adrenergic, beta-1-, receptor; Beta-1 adrenergic receptor; Beta-1 adrenoceptor; Beta-1 adrenoreceptor
Gene Aliases: ADRB1; ADRB1R; B1AR; BETA1AR; RHR
UniProt ID: (Human) P08588
Entrez Gene ID: (Human) 153
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.