Abnova
Manufacturer Code:PAB9919
Catalog # 89-134-493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of drosophila aub. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of aub.
For Research Use Only
Protein Aliases: Aub; aub-PA; aub-PC; CG6137; CG6137-PA; CG6137-PC; ms(2)32D; Protein aubergine; Protein sting; stellate-interacting gene; sti; sting
Gene Aliases: AUB; CG6137; Dmel\CG6137; Dmel_CG6137; ms(2)32D; sti; sting
UniProt ID: (Fruit fly) O76922
Entrez Gene ID: (Fruit fly) 34524
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.