Novus Biologicals
Manufacturer Code:NBP298076V
Catalog # NB052200
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human astacin-like metalloendopeptidase fragment. |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: astacin-like metallo-endopeptidase (M12 family); Astacin-like metalloendopeptidase; astacin-like metalloendopeptidase (M12 family); ASTL; Oocyte astacin; Ovastacin; sperm acrosomal SLLP1 binding; ZP2-proteinase
Gene Aliases: ASTL; SAS1B
UniProt ID: (Human) Q6HA08
Entrez Gene ID: (Human) 431705
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.