R&D Systems
Manufacturer Code:AF8549
Catalog # AF8549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Ovastacin Ala24-Asp431 Accession # Q6HA08 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 to -70 degreesC under sterile conditions |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Astacin-like metalloendopeptidase; astacin-like metalloendopeptidase (M12 family); Astacin-Like Metalloendopeptidase astacin-like metallo-endopeptidase (M12 family) ASTL Oocyte Astacin; ASTL; Oocyte astacin; Ovastacin; sperm acrosomal SLLP1 binding; ZP2-proteinase
Gene Aliases: ASTL; SAS1B
UniProt ID: (Human) Q6HA08
Entrez Gene ID: (Human) 431705
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.