Abnova
Manufacturer Code:H00056052A01
Catalog # 89-004-679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALG1 (NP_061982 154 a.a. ~ 253 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ALG1.
For Research Use Only
Protein Aliases: asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; Asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; Beta-1,4-mannosyltransferase; Chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; HMAT1; HMT-1; HMT1; Mannosyltransferase-1; MT-1
Gene Aliases: ALG1; CDG1K; hMat-1; HMAT1; HMT-1; HMT1; Mat-1; MT-1; PSEC0061; UNQ861/PRO1870
UniProt ID: (Human) Q9BT22
Entrez Gene ID: (Human) 56052
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.