Abnova
Manufacturer Code:H00000242A01
Catalog # 89-004-014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALOX12B (NP_001130 171 a.a. ~ 261 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ALOX12B.
For Research Use Only
Protein Aliases: 12R-lipoxygenase; 12R-LOX; Arachidonate 12-lipoxygenase, 12R-type; Epidermis-type lipoxygenase 12
Gene Aliases: 12R-LOX; ALOX12B; ARCI2
UniProt ID: (Human) O75342
Entrez Gene ID: (Human) 242
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.