Abnova
Manufacturer Code:H00026057M02
Catalog # 89-001-239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B8 |
Immunogen | ANKRD17 (NP_115593 2501 a.a. ~ 2603 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ANKRD17.
For Research Use Only
Protein Aliases: Ankyrin repeat domain-containing protein 17; FLJ22206; Gene trap ankyrin repeat protein; GTAR; KIAA0697; NY-BR-16; Serologically defined breast cancer antigen NY-BR-16
Gene Aliases: ANKRD17; GTAR; KIAA0697; MASK2; NY-BR-16
UniProt ID: (Human) Q96I86
Entrez Gene ID: (Human) 26057
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.