Abnova
Manufacturer Code:H00000277B01
Catalog # 89-004-893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | AMY1B (NP_001008219.1 1 a.a. ~ 511 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human AMY1B protein.
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; AMY1; amylase, salivary, alpha-1B; glycogenase; Salivary alpha-amylase; salivary amylase alpha 1B
Gene Aliases: AMY1; AMY1A; AMY1B; AMY1C
UniProt ID: (Human) P0DUB6
Entrez Gene ID: (Human) 277
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.