Invitrogen
Manufacturer Code:MA119162
Catalog # 01-675-932
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TU-01 |
Immunogen | Fraction of tubulin purified from porcine brain by two cycles of polymerization - depolymerization. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-tubulin 1; Alpha-tubulin 4; Alpha-tubulin isotype M-alpha-4; FLJ30169 H2 ALPHA Testis specific alpha tubulin TUBA1 Tubulin alpha; Testis-specific alpha-tubulin; Tubulin alpha-1 chain; Tubulin alpha-4 chain; Tubulin alpha-4A chain; Tubulin H2-alpha; tubulin, alpha 1; tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a
Gene Aliases: ALS22; H2-ALPHA; M[a]4; TUBA1; Tuba4; TUBA4A
UniProt ID: (Human) B3KNQ6, (Bovine) P81948, (Rat) Q5XIF6, (Mouse) P68368
Entrez Gene ID: (Human) 7277, (Bovine) 777775, (Rat) 316531, (Mouse) 22145
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.