Novus Biologicals
Manufacturer Code:NB100690SS
Catalog # NB100690SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | DM1A |
Immunogen | This alpha Tubulin Antibody (DM1A) was developed against native chicken brain microtubules. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha tubulin loading control alpha tubulin monoclonal Alpha-tubulin 3 B-ALPHA-1 DM1A DM1A anti-tubulin dm1a monoclonal DM1a tubulin FLJ25113 hum-a-tub1 hum-a-tub2 LIS3 TUBA1 monoclonal TUBA3 tubulin alpha-1A chain Tubulin alpha-3 chain Tubulin B-alpha-1 tubulin monoclonal tubulin alpha 1a tubulin alpha 3 tubulin alpha brain-specific; Alpha-tubulin 3; Detyrosinated tubulin alpha-1A chain; hum-a-tub1; hum-a-tub2; Tubulin alpha-1A chain; Tubulin alpha-3 chain; Tubulin B-alpha-1; tubulin, alpha 1a; tubulin, alpha, brain-specific
Gene Aliases: B-ALPHA-1; LIS3; TUBA1A; TUBA3
UniProt ID: (Human) Q71U36
Entrez Gene ID: (Human) 7846
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.