Invitrogen
Manufacturer Code:PA529444
Catalog # PIPA529444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 154 and 448 of human alpha Tubulin 4a |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: &agr-tubulin 84B; alpha tubulin; alpha tubulin 84B; alpha-Tubulin; Alpha-tubulin 1; Alpha-tubulin 4; alpha-Tubulin 84B; Alpha-tubulin isotype M-alpha-4; alpha-tubulin84B; alpha1 tubulin; alpha1-tubulin; alpha[[1]] tubulin; alpha[[1]]-tubulin; alphaT; alphaTub84B-PA; alphatubulin; alphaTubulin 84B; alphaTubulin84B; AT25469p; CG1913-PA; H2-ALPHA TUBA1 tubulin H2-alpha tubulin alpha-1 chain tubulin alpha-4A chain tubulin alpha 1 (testis specific); Testis-specific alpha-tubulin; tubulin; tubulin 1 alpha-chain; tubulin 1alpha; Tubulin alpha-1 chain; Tubulin alpha-4 chain; Tubulin alpha-4A chain; Tubulin alpha-5 chain; tubulin alpha1; tubulin alpha1 promoter; Tubulin H2-alpha; tubulin(alpha1); tubulin, alpha 1 (testis specific); tubulin, alpha 4; tubulin, alpha 4a; Tubulin-alpha1; tubulin1alpha; tubulin84B
Gene Aliases: 1t; a-tub84B; a1t; ALPHA 84C; alpha-Tub; alpha-Tub84B; alpha-tubulin; alpha-tubulin 84B; Alpha-Tubulin84B; alpha1; alpha1t; alpha1tub; alpha1tub84B; alpha84B; alphaT; alphat-1; alphaTUB; alphaTub1; alphaTub84; alphaTUB84B; ALS22; anon-EST:Liang-1.59; anon-EST:Liang-2.30; aTub84B; BEST:LD32507; CG1913; chr3R:2914276..2914416; clone 1.59; clone 2.30; DM1alpha; Dmel\CG1913; Dmel_CG1913; DTA1; H2-ALPHA; l(3)84Bd; l(3)g3; M[a]4; ms(3)nc33; nc33; T; Talpha1; TBA1_DROME; Tub; Tub 84B; Tub1C; Tub84B; TUBA1; Tuba4; TUBA4A; Tuba84B; Tubalpha1; tubulin
UniProt ID: (Human) B3KNQ6, (Rat) Q5XIF6, (Mouse) P68368, (Fruit fly) P06603
Entrez Gene ID: (Human) 7277, (Chicken) 100857247, (Rat) 316531, (Mouse) 22145, (Fruit fly) 40848
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.