Invitrogen
Manufacturer Code:PA522060
Catalog # PIPA522060
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment contain a sequence corresponding to a region within amino acids 37 and 276 (Q71U36) of alpha Tubulin 1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: &agr-tubulin 84B; alpha tubulin; alpha tubulin 84B; alpha-Tubulin; Alpha-tubulin 1; Alpha-tubulin 3; alpha-Tubulin 84B; Alpha-tubulin isotype M-alpha-1; alpha-tubulin84B; alpha1 tubulin; alpha1-tubulin; alpha[[1]] tubulin; alpha[[1]]-tubulin; alphaT; alphaTub84B-PA; alphatubulin; alphaTubulin 84B; alphaTubulin84B; AT25469p; B-ALPHA-1 LIS3 TUBA3 alpha-tubulin 3 hum-a-tub1 hum-a-tub2 tubulin B-alpha-1 tubulin alpha-1A chain tubulin alpha-3 chain tubulin alpha 3 tubulin alpha brain-specific; CG1913-PA; Detyrosinated tubulin alpha-1A chain; hum-a-tub1; hum-a-tub2; Talpha1 alpha-tubulin; tubulin; tubulin 1 alpha-chain; tubulin 1alpha; Tubulin alpha-1 chain; Tubulin alpha-1A chain; Tubulin alpha-3 chain; tubulin alpha1; tubulin alpha1 promoter; Tubulin B-alpha-1; tubulin(alpha1); tubulin, alpha 1; tubulin, alpha 1a; tubulin, alpha, brain-specific; Tubulin-alpha1; tubulin1alpha; tubulin84B
Gene Aliases: 1t; a-tub84B; a1t; ALPHA 84C; alpha-Tub; alpha-Tub84B; alpha-tubulin; alpha-tubulin 84B; Alpha-Tubulin84B; alpha1; alpha1t; alpha1tub; alpha1tub84B; alpha84B; alphaT; alphat-1; alphaTUB; alphaTub1; alphaTub84; alphaTUB84B; anon-EST:Liang-1.59; anon-EST:Liang-2.30; aTub84B; B-ALPHA-1; BEST:LD32507; CG1913; chr3R:2914276..2914416; clone 1.59; clone 2.30; DM1alpha; Dmel\CG1913; Dmel_CG1913; DTA1; l(3)84Bd; l(3)g3; LIS3; ms(3)nc33; nc33; T; Talpha1; TBA1_DROME; Tub; Tub 84B; Tub1C; Tub84B; Tuba-1; Tuba1; TUBA1A; TUBA3; Tuba84B; Tubalpha1; tubulin
UniProt ID: (Human) Q71U36, (Mouse) P68369, (Rat) P68370, (Fruit fly) P06603
Entrez Gene ID: (Human) 7846, (Mouse) 22142, (Rat) 64158, (Fruit fly) 40848
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein tubulin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.