Invitrogen
Manufacturer Code:PA589852
Catalog # PIPA589852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-110 of human TTPA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha TTP; Alpha-tocopherol transfer protein; Alpha-TTP; Tocopherol transfer protein alpha
Gene Aliases: alpha-TTP; Tpp1; TTP; Ttpa
UniProt ID: (Rat) P41034, (Mouse) Q8BWP5
Entrez Gene ID: (Rat) 25571, (Mouse) 50500
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.