Invitrogen
Manufacturer Code:PA513401
Catalog # PIPA513401
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 104-133 amino acids from the C-terminal region of human SNCA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha synuclein antibody MGC110988 antibody NACP antibodyNon A beta component of AD amyloid antibody Non A4 component of amyloid precursor antibody PARK 1 antibody PARK 4 antibody PARK1 antibody PARK4 antibody Parkinson disease familial 1 antibody PD 1 antibody PD1 antibody SNCA antibody Synuclein alpha antibody; Alpha-synuclein; NACP; non A-beta component of AD amyloid; Non-A beta component of AD amyloid; non-A4 component of amyloid; Non-A4 component of amyloid precursor; synuclein alpha-140; synuclein, alpha (non A4 component of amyloid precursor)
Gene Aliases: alphaSYN; NACP; PARK1; PARK4; PD1; SNCA; Syn
UniProt ID: (Human) P37840, (Mouse) Q9CXF8
Entrez Gene ID: (Human) 6622, (Mouse) 20617
Molecular Function: chaperone membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.