Novus Biologicals
Manufacturer Code:NBP278836
Catalog # NB002998
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | BLR082G |
Immunogen | Alpha-Smooth Muscle Actin Antibody (BLR082G) is a recombinant monoclonal made to a peptide representing a region between residue between 1 and 50 of human aortic smooth muscle actin using the numbering given in entry NP_001135417.1 (Gene ID 59). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Monoclonal Antibody
Protein Aliases: 1a4 sma 1a4 smooth muscle actin AAT6 actin alpha 2 smooth muscle aorta actin aortic smooth muscle ACTSA ACTVS alpha 2 actin alpha SMA alpha sma 1a4 alpha smooth muscle actin alpha-actin-2 alpha-SMA anti-alpha SMA a-sma Cell growth-inhibiting gene 46 protein growth-inhibiting gene 46 sma sma 1a4 smooth muscle actin; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; Alpha-actin-2; alpha-cardiac actin; Cell growth-inhibiting gene 46 protein
Gene Aliases: AAT6; ACTA2; ACTSA; ACTVS; GIG46; MYMY5
UniProt ID: (Human) P62736
Entrez Gene ID: (Human) 59
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.