Novus Biologicals
Manufacturer Code:NBP237641
Catalog # NBP237641
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1H8 |
Immunogen | Alpha-Smooth Muscle Actin Antibody (1H8) is made to a purified recombinant fragment of human ACTA2 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AAT6 actin alpha 2 smooth muscle aorta actin aortic smooth muscle ACTSA ACTVS alpha 2 actin alpha SMA alpha smooth muscle actin alpha-actin-2 alpha-SMA a-sma Cell growth-inhibiting gene 46 protein growth-inhibiting gene 46 sma smooth muscle actin; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; Alpha-actin-2; alpha-cardiac actin; Cell growth-inhibiting gene 46 protein
Gene Aliases: AAT6; ACTA2; ACTSA; ACTVS; GIG46; MYMY5
UniProt ID: (Human) P62736
Entrez Gene ID: (Human) 59
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.