R&D Systems
Manufacturer Code:AF7159SP
Catalog # AF7159SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human alpha-N-terminal Methyltransferase 1A/METTL11A Thr2-Arg223 Accession # Q9BV86 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AD-003 alpha N-terminal protein methyltransferase 1A C9orf32 chromosome 9 open reading frame 32 EC 2.1.1.- methyltransferase like 11A Methyltransferase-like protein 11A NRMT N-terminal RCC1 methyltransferase NTM1A NTMT1 X-Pro-Lys N-terminal protein methyltransferase 1A; Alpha N-terminal protein methyltransferase 1A; Methyltransferase-like protein 11A; N-terminal RCC1 methyltransferase; N-terminal Xaa-Pro-Lys N-methyltransferase 1; N-terminal Xaa-Pro-Lys N-methyltransferase 1, N-terminally processed; NTM1A; X-Pro-Lys N-terminal protein methyltransferase 1A
Gene Aliases: AD-003; C9orf32; HOMT1A; METTL11A; NRMT; NRMT1; NTM1A; NTMT1
UniProt ID: (Human) Q5SZB9
Entrez Gene ID: (Human) 28989
Molecular Function:
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.