Novus Biologicals
Manufacturer Code:NBP300180G
Catalog # NB064752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant alpha-N-acetylgalactosaminidase/NAGA (NP_000253.1 Met1-Gln411). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); Alpha-galactosidase B; alpha-N- (alpha-galactosidase B) alpha-N-acetylgalactosaminidase EC 3.2.1 EC 3.2.1.49 GALB N-acetylgalactosaminidase alpha-; Alpha-N-acetylgalactosaminidase; N-acetylgalactosaminidase, alpha-
Gene Aliases: D22S674; GALB; NAGA
UniProt ID: (Human) P17050
Entrez Gene ID: (Human) 4668
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.