R&D Systems
Manufacturer Code:AF6717
Catalog # AF6717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line recombinant human alpha-N-acetylgalactosaminidase/NAGA Leu18-Gln411 Accession # P17050 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); Alpha-galactosidase B; alpha-N- (alpha-galactosidase B) alphaNacetylgalactosaminidase alpha-N-acetylgalactosaminidase EC 3.2.1 EC 3.2.1.49 GALB N-acetylgalactosaminidase alpha- NAGA; Alpha-N-acetylgalactosaminidase; N-acetylgalactosaminidase, alpha-
Gene Aliases: D22S674; GALB; NAGA
UniProt ID: (Human) P17050
Entrez Gene ID: (Human) 4668
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.