Novus Biologicals
Manufacturer Code:NBP288931
Catalog # NB036521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL9362 |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: APFYTTYRTADGEFMAVGAIEPQFYELLIKGLGLKSDELPNQMSMDDWPEMKKKFADVFAKKTKAEWCQIFDGTDACVTPVLTF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°:C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2-methylacyl-CoA racemase; 2-methylacyl-CoA racemase alpha-methylacyl-CoA racemase CBAS4 EC 5.1.99.4 RACE RM; Alpha-methylacyl-CoA racemase
Gene Aliases: AMACR; AMACRD; CBAS4; RACE; RM
UniProt ID: (Human) Q9UHK6
Entrez Gene ID: (Human) 23600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.