R&D Systems
Manufacturer Code:MAB4844SP
Catalog # MAB4844SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 529724 |
Immunogen | <EM>E. coli</EM>-derived recombinant human alpha-Internexin Val230-Glu450 Accession # Q16352 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 66 kDa neurofilament protein; 66 kDa neurofilament protein alphaInternexin alpha-Inx FLJ18662 FLJ57501 INA internexin neuronal intermediate filament protein alpha MGC12702 NEF5 Neurofilament 5 neurofilament 5 (66kD) Neurofilament-66 neurofilament-66 tax-binding protein NF-66 NF-66alpha-internexin TXBP-1; Alpha-internexin; Alpha-Inx; internexin neuronal intermediate filament protein, alpha; Neurofilament 5; neurofilament 5 (66kD); neurofilament-66, tax-binding protein; NF-66
Gene Aliases: INA; NEF5; NF-66; TXBP-1
UniProt ID: (Human) Q16352
Entrez Gene ID: (Human) 9118
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.