Novus Biologicals
Manufacturer Code:NBP229978
Catalog # NBP229978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 257CT7.1.2 |
Immunogen | This INA antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 290-319 amino acids from human INA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 66 kDa neurofilament protein; 66 kDa neurofilament protein alpha-Inx FLJ18662 FLJ57501 internexin neuronal intermediate filament protein alpha MGC12702 NEF5 Neurofilament 5 neurofilament 5 (66kD) Neurofilament-66 neurofilament-66 tax-binding protein NF-66alpha-internexin TXBP-1; Alpha-internexin; Alpha-Inx; internexin neuronal intermediate filament protein, alpha; Neurofilament 5; neurofilament 5 (66kD); neurofilament-66, tax-binding protein; NF-66
Gene Aliases: INA; NEF5; NF-66; TXBP-1
UniProt ID: (Human) Q16352
Entrez Gene ID: (Human) 9118
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.