Invitrogen
Manufacturer Code:MA316752
Catalog # AMSA6752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2 |
Immunogen | Raised against purified recombit rat alpha-internexin expressed in and purified from E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 66 kDa neurofilament protein; Alpha-internexin; Alpha-Inx; FLJ18662 FLJ57501 MGC12702 NEF5 NF-66 TXBP-1; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Neurofilament 5; neurofilament 5 (66kD); neurofilament-66; neurofilament-66, tax-binding protein; NF-66
Gene Aliases: AV028420; INA; Inexa; Intlaa; NEF5; NF-66; NF66; TXBP-1
UniProt ID: (Human) Q16352, (Bovine) Q08DH7, (Mouse) P46660
Entrez Gene ID: (Human) 9118, (Bovine) 532236, (Mouse) 226180, (Rat) 24503
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.