Invitrogen
Manufacturer Code:PA580737
Catalog # PIPA580737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the C-terminus of the Human INA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 66 kDa neurofilament protein; 66 kDa neurofilament protein Internexin neuronal intermediate filament protein alpha MGC12702 NEF5 Neurofilament 5 (66kD) Neurofilament 66 Neuro; Alpha-internexin; Alpha-Inx; internexin neuronal intermediate filament protein, alpha; Neurofilament 5; neurofilament 5 (66kD); neurofilament-66, tax-binding protein; NF-66
Gene Aliases: INA; NEF5; NF-66; TXBP-1
UniProt ID: (Human) Q16352
Entrez Gene ID: (Human) 9118
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.