Invitrogen
Manufacturer Code:PA110017
Catalog # 01-670-274
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Purified recombit rat alpha-internexin expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 66 kDa neurofilament protein; Alpha-internexin; Alpha-Inx; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Neurofilament 5; neurofilament 5 (66kD); neurofilament-66; neurofilament-66, tax-binding protein; NF-66; NF66 alpha-Inx 66 kDa neurofilament protein Neurofilament-66
Gene Aliases: AV028420; INA; Inexa; Intlaa; NEF5; NF-66; NF66; TXBP-1
UniProt ID: (Human) Q16352, (Bovine) Q08DH7, (Mouse) P46660
Entrez Gene ID: (Human) 9118, (Pig) 100153400, (Horse) 100059769, (Bovine) 532236, (Mouse) 226180, (Rat) 24503
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.