Invitrogen
Manufacturer Code:PA544905
Catalog # PIPA544905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SPTAN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a--fodrin; alpha II spectrin; alpha-fodrin; Alpha-II spectrin; alpha-spectrin 2; alpha-spectrin 2, brain; alpha2 spectrin; brain alpha-spectrin; cardiomyocyte alpha-spectrin; Fodrin alpha chain; inhibitory protein factor; noerythroid alpha-spectrin 2; nonerythroid alpha-spectrin 2; sequence with a GC signal; spectrin alpha 2; spectrin alpha chain, brain; Spectrin alpha chain, non-erythrocytic 1; Spectrin alpha non-erythrocytic 1 (alpha-fodrin) (ALPHA)II-SPECTRIN FLJ44613 NEAS EIEE5 SPTA2; spectrin, alpha, non-erythrocytic 1 (alpha-fodrin); spectrin, non-erythroid alpha chain; Spectrin, non-erythroid alpha subunit
Gene Aliases: 2610027H02Rik; A2a; EIEE5; im:7157190; IPF; NEAS; Spna-2; Spna2; SPTA2; SPTAN1; wu:fa20e05; wu:fb33g08; wu:fk32a10; zgc:112229
UniProt ID: (Human) Q13813, (Rat) P16086
Entrez Gene ID: (Horse) 100066945, (Dog) 403484, (Rabbit) 100350866, (Bovine) 540108, (Pig) 100049693, (Human) 6709, (Mouse) 20740, (Rat) 64159, (Zebrafish) 553383
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.