R&D Systems
Manufacturer Code:MAB1368SP
Catalog # MAB1368SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 189502 |
Immunogen | Human umbilical cord serum-derived alpha-Fetoprotein/AFP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AFP Alpha-1-fetoprotein Alpha-fetoglobulin alpha-fetoprotein DSCAM2 FETA HP HPAFP; Alpha-1-fetoprotein; Alpha-fetoglobulin; Alpha-fetoprotein
Gene Aliases: AFP; AFPD; FETA; HPAFP
UniProt ID: (Human) P02771
Entrez Gene ID: (Human) 174
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.