Invitrogen
Manufacturer Code:PA531516
Catalog # PIPA531516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 150 and 379 of Human alpha amylase 2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 2B; Alpha-amylase 2B; AMY2 14-alpha-D-glucan glucanohydrolase 2B alpha-amylase 2B alpha-amylase carcinoid carcinoid alpha-amylase glycogenase; Carcinoid alpha-amylase; glycogenase
Gene Aliases: AMY2; AMY2B; AMY3; HXA
UniProt ID: (Human) P19961
Entrez Gene ID: (Human) 280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.