Invitrogen
Manufacturer Code:PA570037
Catalog # PIPA570037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat alpha Amylase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; alpha-amylase 1; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; AMY1 AMY1A AMY1B 14-alpha-D-glucan glucanohydrolase 1 alpha-amylase 1 amylase salivary alpha-1C glycogenase salivary alpha-amylase salivary amylase alpha 1C; amylase 1, salivary; amylase, salivary, alpha-1A; glycogenase; Salivary alpha-amylase; salivary amylase alpha 1A
Gene Aliases: AMY1; AMY1A; AMY1B; AMY1C
UniProt ID: (Human) P0DUB6
Entrez Gene ID: (Human) 276, (Rat) 24203
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.