Invitrogen
Manufacturer Code:PA5111216
Catalog # PIPA5111216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human alpha Amylase 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; 14-alpha-D-glucan glucanohydrolase 1 Alpha-amylase 1 AMY1 Amy-1 AMY1A Amy-1-a AMY1B AMY1C AMY2A C030014B17Rik PA Salivary alpha-amylase Salivary and hepatic alpha-amylase; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; amylase, salivary, alpha-1A; glycogenase; Salivary alpha-amylase; salivary amylase alpha 1A
Gene Aliases: AMY1; AMY1A; AMY1B; AMY1C
UniProt ID: (Human) P0DUB6
Entrez Gene ID: (Human) 276
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.