Proteintech
Manufacturer Code:114011AP150UL
Catalog # 50-172-7127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | alpha Adaptin Fusion Protein Ag1927 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 100 kDa coated vesicle protein A; adapter-related protein complex 2 alpha-1 subunit; adapter-related protein complex 2 subunit alpha-1; adaptin, alpha A; Adaptor protein complex AP-2 subunit alpha-1; adaptor protein complex AP-2, alpha 1 subunit; adaptor related protein complex 2, alpha 1 subunit; Adaptor-related protein complex 2 subunit alpha-1; adaptor-related protein complex 2, alpha 1 subunit; adaptor-related protein complex AP-2, alpha 1 subunit; ADTAA alpha Adaptin Alpha adaptin A Alpha1 adaptin AP 2 complex subunit alpha 1 AP2 ALPHA AP2A1 CLAPA1; Alpha-adaptin A; Alpha1-adaptin; AP-2 complex subunit alpha-1; Clathrin assembly protein complex 2 alpha-A large chain; clathrin-associated/assembly/adaptor protein, large, alpha 1; Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit
Gene Aliases: ADTAA; AP2-ALPHA; AP2A1; CLAPA1
UniProt ID: (Human) O95782, (Mouse) P17426
Entrez Gene ID: (Human) 160, (Rat) 308578, (Mouse) 11771
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.