Invitrogen
Manufacturer Code:MA536095
Catalog # PIMA536095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | A1G2 |
Immunogen | Synthetic peptide within human ACTN1 aa 350-650. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3110023F10Rik actinin 1 smooth muscle actinin alpha 1 actinin alpha 1 ACTN1 Actn1a alpha actinin 1a alpha-actinin cytoskeletal isoform Alpha-actinin-1 BDPLT15 F-actin cross-linking protein FLJ40884 FLJ54432 I79_004057 non-muscle alpha-actinin 1 non-muscle alpha-actinin-1 RCJMB04_23c5; actinin 1 smooth muscle; actinin, alpha 1; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; F-actin cross-linking protein; Non-muscle alpha-actinin-1
Gene Aliases: ACTN1; BDPLT15
UniProt ID: (Human) P12814
Entrez Gene ID: (Human) 87
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.