Invitrogen
Manufacturer Code:PA514898
Catalog # PIPA514898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 192-220 amino acids from the N-terminal region of human ACTN4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-actinin-4; Alpha-actinin-4 Non-muscle alpha-actinin 4 F-actin cross-linking protein ACTN4; focal segmental glomerulosclerosis 1; Non-muscle alpha-actinin 4
Gene Aliases: ACTININ-4; ACTN4; FSGS; FSGS1
UniProt ID: (Human) O43707
Entrez Gene ID: (Human) 81
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.