Invitrogen
Manufacturer Code:MA533007
Catalog # PIMA533007
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 9B5 |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human ACTN3 different from the related mouse sequence by five amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-3; Alpha-actinin-3 Alpha-actinin skeletal muscle isoform 3 F-actin cross-linking protein ACTN3; F-actin cross-linking protein
Gene Aliases: Actn3
UniProt ID: (Mouse) O88990
Entrez Gene ID: (Mouse) 11474, (Rat) 171009
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.