Invitrogen
Manufacturer Code:PA599425
Catalog # PIPA599425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Actinin alpha 2/3 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110008F24Rik actinin alpha 2 actinin alpha 3 actinin alpha 3 (gene/pseudogene) Actinin alpha3 actinin alpha 2 actinin alpha 3 Actinin-alpha3 Actn2 ACTN3 alpha-actinin skeletal muscle Alpha-actinin skeletal muscle isoform 2 Alpha-actinin skeletal muscle isoform 3 Alpha-actinin-2 alpha-Actinin3 Alpha-actinin-3 alphaActinin-3 CMD1AA CMH23 F-actin cross-linking protein; actinin, alpha 2; actinin, alpha 3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-2; Alpha-actinin-3; F-actin cross-linking protein
Gene Aliases: 1110008F24Rik; ACTN2; ACTN3; CMD1AA; CMH23
UniProt ID: (Human) P35609, (Human) Q08043, (Mouse) Q9JI91, (Mouse) O88990
Entrez Gene ID: (Human) 88, (Human) 89, (Mouse) 11472, (Rat) 291245, (Mouse) 11474, (Rat) 171009
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.