Invitrogen
Manufacturer Code:PA538605
Catalog # PIPA538605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the N-terminal region of human Actin alpha-2/3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actinin, alpha 2; actinin, alpha 3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-2; Alpha-actinin-3; CMD1AA F-actin cross-linking protein alpha-actinin skeletal muscle alpha-actinin-2 Alpha-actinin-3 Alpha-actinin skeletal muscle isoform 3 ACTN3 alpha-Actinin3 Actinin alpha3 Actinin alpha3 alphaActinin-3 Actinin-alpha3 alpha-actinin-3; F-actin cross-linking protein
Gene Aliases: 1110008F24Rik; ACTN2; ACTN3; CMD1AA; CMH23
UniProt ID: (Human) P35609, (Human) Q08043, (Mouse) Q9JI91, (Mouse) O88990
Entrez Gene ID: (Human) 88, (Human) 89, (Mouse) 11472, (Rat) 291245, (Mouse) 11474, (Rat) 171009
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.