Novus Biologicals
Manufacturer Code:NBP270078AF594
Catalog # NP27078A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7A4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 650-893 of human alpha-Actinin 1 (NP_001093) produced in E.coli. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: actinin 1 smooth muscle; actinin 1 smooth muscle actinin alpha 1 alpha-actinin 1 Alpha-actinin cytoskeletal isoform alpha-actinin-1 F-actin cross-linking protein FLJ40884 FLJ54432 Non-muscle alpha-actinin-1; actinin, alpha 1; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; F-actin cross-linking protein; Non-muscle alpha-actinin-1
Gene Aliases: ACTN1; BDPLT15
UniProt ID: (Human) P12814
Entrez Gene ID: (Human) 87
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.