R&D Systems
Manufacturer Code:MAB8279
Catalog # MAB8279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 910301 |
Immunogen | <EM>E. coli</EM>-derived recombinant human alpha-Actinin 1 Asn272-Thr500 Accession # P12814 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: actinin 1 smooth muscle; actinin 1 smooth muscle actinin alpha 1 ACTN1 alphaActinin 1 alpha-Actinin 1 Alpha-actinin cytoskeletal isoform alpha-actinin-1 BDPLT15 F-actin cross-linking protein FLJ40884 FLJ54432 Non-muscle alpha-actinin-1; actinin, alpha 1; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; F-actin cross-linking protein; Non-muscle alpha-actinin-1
Gene Aliases: ACTN1; BDPLT15
UniProt ID: (Human) P12814
Entrez Gene ID: (Human) 87
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.