Invitrogen
Manufacturer Code:PA517308
Catalog # PIPA517308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to carboxy-terminal residues of human a-actinin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actinin 1 smooth muscle; actinin, alpha 1; alpha actinin 1a; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; F-actin cross-linking protein; F-actin cross-linking protein actinin 1 smooth muscle alpha-actinin 1 alpha-actinin cytoskeletal isoform alpha-actinin-1 non-muscle alpha-actinin-1; non-muscle alpha-actinin 1; Non-muscle alpha-actinin-1
Gene Aliases: 3110023F10Rik; ACTN1; Actn1a; BDPLT15; I79_004057
UniProt ID: (Human) P12814, (Rat) Q9Z1P2, (Mouse) Q7TPR4
Entrez Gene ID: (Human) 87, (Rat) 81634, (Mouse) 109711, (Chinese hamster) 100755831
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.