Novus Biologicals
Manufacturer Code:NBP289158
Catalog # NB036741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein within human alpha Actinin aa 350-650. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actinin 1 smooth muscle; actinin 1 smooth muscle actinin alpha 1 alpha-actinin 1 Alpha-actinin cytoskeletal isoform alpha-actinin-1 F-actin cross-linking protein FLJ40884 FLJ54432 Non-muscle alpha-actinin-1; actinin, alpha 1; Alpha-actinin cytoskeletal isoform; Alpha-actinin-1; F-actin cross-linking protein; Non-muscle alpha-actinin-1
Gene Aliases: ACTN1; BDPLT15
UniProt ID: (Human) P12814
Entrez Gene ID: (Human) 87
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.